7-Bromo-1,2,3,4-tetrahydroquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR301354-1G OR301354-5G
APOSOR301354-1GEA 414 DKK
APOSOR301354-1G APOSOR301354-5G
7-Bromo-1,2,3,4-tetrahydroquinoline
7-Bromo-1,2,3,4-tetrahydroquinoline
Formel: C₉H₁₀BrN
Molekylvægt: 212.1 g/mol
Smeltepunkt: 68…69 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08544270
CAS-nummer: 114744-51-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR