6-Bromochroman-4-one 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR301108-5G OR301108-100G OR301108-25G
APOSOR301108-5GEA 183 DKK
APOSOR301108-5G APOSOR301108-100G APOSOR301108-25G
6-Bromochroman-4-one 98%
6-Bromochroman-4-one
Formel: C₉H₇BrO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00546803
CAS-nummer: 49660-57-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR