2-Methyl-2-phenylpropanenitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR29220-1G
OR29220-250MG
OR29220-5G
APOSOR29220-1GEA
676
DKK
APOSOR29220-1G
APOSOR29220-250MG
APOSOR29220-5G
2-Methyl-2-phenylpropanenitrile
2-Methyl-2-phenylpropanenitrile
Formel:
C₁₀H₁₁N Kogepunkt: 54…56 °C/1mmHg Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00099614 CAS-nummer: 1195-98-8 UN nummer: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...