1-Ethylimidazol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR2905-100G
OR2905-500G
APOSOR2905-100GEA
164
DKK
APOSOR2905-100G
APOSOR2905-500G
1-Ethylimidazol
1-Ethylimidazole
Formel:
C₅H₈N₂ Kogepunkt: 104 °C/262mmHg Smeltepunkt: –27 °C Massefylde: 0,995 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00055389 CAS-nummer: 7098-07-9 EINECS: 230-403-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...