3-Amino-4-hydroxy-5-nitrobenzene sulphonic acid monohydrate
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
OR28927-10G OR28927-1G
APOSOR28927-10GEA 382 DKK
APOSOR28927-10G APOSOR28927-1G
3-Amino-4-hydroxy-5-nitrobenzene sulphonic acid monohydrate
3-Amino-4-hydroxy-5-nitrobenzene sulphonic acid monohydrate
Formel: C₆H₆N₂O₆S·1H₂O
Smeltepunkt: 300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00277203
CAS-nummer: 175278-60-1