Aminomalononitrile-p-toluenesulphonate 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Aminomalononitrile p-Toluenesulphonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Dicyanomethylammonium p-Toluenesulfonate
OR28794-10G
OR28794-25G
OR28794-5G
APOSOR28794-10GEA
382
DKK
APOSOR28794-10G
APOSOR28794-25G
APOSOR28794-5G
Aminomalononitrile-p-toluenesulphonate 97%
Aminomalononitrile-p-toluenesulphonate
Formel:
C₃H₃N₃ Molekylvægt: 253.28 g/mol Smeltepunkt: 169 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00012848 CAS-nummer: 5098-14-6 EINECS: 225-817-1 UN nummer: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...