2-Isopropylanilin 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR28577-100G
OR28577-500G
OR28577-5G
APOSOR28577-100GEA
850
DKK
APOSOR28577-100G
APOSOR28577-500G
APOSOR28577-5G
2-Isopropylanilin 97%
2-Isopropylanilin
Formel:
C₉H₁₃N Kogepunkt: 214…216 °C/10mmHg Massefylde: 0,966 g/cm³ (25 °C) Flammepunkt: 95 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007720 CAS-nummer: 643-28-7 EINECS: 211-397-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...