5-Chloro-6-hydroxynicotinic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR2842-25G OR2842-100G
APOSOR2842-25GEA 387 DKK
APOSOR2842-25G APOSOR2842-100G
5-Chloro-6-hydroxynicotinic acid 98%
5-Chloro-6-hydroxynicotinic acid
Formel: C₆H₄ClNO₃
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052950
CAS-nummer: 54127-63-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR