Thiophene-2,5-dicarbaldehyde
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR28138-1G
OR28138-5G
APOSOR28138-1GEA
183
DKK
APOSOR28138-1G
APOSOR28138-5G
Thiophene-2,5-dicarbaldehyde
Thiophene-2,5-dicarbaldehyde
Formel:
C₆H₄O₂S Smeltepunkt: 114…117 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00216592 CAS-nummer: 932-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...