4-Amino-6-chloro-2-(methylthio)pyrimidine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Amino-6-chloro-2-methylmercaptopyrimidine

OR27855-100G OR27855-25G
APOSOR27855-100GEA 1780 DKK
APOSOR27855-100G APOSOR27855-25G
4-Amino-6-chloro-2-(methylthio)pyrimidine 95%
4-Amino-6-chloro-2-(methylthio)pyrimidine
Formel: C₅H₆ClN₃S
Molekylvægt: 175.6400 g/mol
Smeltepunkt: 130…132 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006088
CAS-nummer: 1005-38-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR