1-Brom-4-chlor-2-nitrobenzen

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR27686-100G OR27686-1KG OR27686-500G
APOSOR27686-100GEA 491 DKK
APOSOR27686-100G APOSOR27686-1KG APOSOR27686-500G
1-Brom-4-chlor-2-nitrobenzen
1-Brom-4-chlor-2-nitrobenzen
Formel: C₆H₃BrClNO₂
Smeltepunkt: 67…72 °C
Massefylde: 2,048 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00024320
CAS-nummer: 41513-04-6
EINECS: 255-421-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR