Methyl-4-amino-3-chloro-5-nitrobenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 4-amino-3-chloro-5-nitrobenzoate

OR2752-1G OR2752-5G
APOSOR2752-1GEA 164 DKK
APOSOR2752-1G APOSOR2752-5G
Methyl-4-amino-3-chloro-5-nitrobenzoate
Methyl-4-amino-3-chloro-5-nitrobenzoate
Formel: C₈H₇ClN₂O₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09842615
CAS-nummer: 863886-04-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR