4-tert-Butyl-o-phenylendiamin
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-(tert-Butyl)benzene-1,2-diamine, 4-tert-Butylbenzene-1,2-diamine, 4-tert-Butyl-1,2-phenylenediamine, 1,2-Diamino-4-(dimethylethyl)benzene, 1,2-Diamino-4-tert-butylbenzene
OR27181-1G
OR27181-250MG
OR27181-5G
APOSOR27181-1GEA
218
DKK
APOSOR27181-1G
APOSOR27181-250MG
APOSOR27181-5G
4-tert-Butyl-o-phenylendiamin
4-tert-Butyl-o-phenylendiamin
Formel:
C₁₀H₁₆N₂ Molekylvægt: 164.25 g/mol Smeltepunkt: 96…100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00052695 CAS-nummer: 68176-57-8 UN nummer: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...