2-Cyano-5-methylbenzenesulphonyl chloride 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR25992-1G OR25992-250MG OR25992-5G
APOSOR25992-1GEA 2430 DKK
APOSOR25992-1G APOSOR25992-250MG APOSOR25992-5G
2-Cyano-5-methylbenzenesulphonyl chloride 98%
2-Cyano-5-methylbenzenesulphonyl chloride
Formel: C₈H₆ClNO₂S
Smeltepunkt: 67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03659720
CAS-nummer: 197960-31-9
UN nummer: 3261
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR