6-Ethoxypyridine-2-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR2588-1G OR2588-250MG OR2588-500MG
APOSOR2588-1GEA 2620 DKK
APOSOR2588-1G APOSOR2588-250MG APOSOR2588-500MG
6-Ethoxypyridine-2-carboxylic acid
6-Ethoxypyridine-2-carboxylic acid
Formel: C₈H₉NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09414713
CAS-nummer: 42955-22-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR