Methyl5-methylnicotinat

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR25732-1G OR25732-5G
APOSOR25732-1GEA 162 DKK
APOSOR25732-1G APOSOR25732-5G
Methyl5-methylnicotinat
Methyl5-methylnicotinat
Formel: C₈H₉NO₂
Kogepunkt: 90 °C/3mmHg
Smeltepunkt: 42…46 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01075739
CAS-nummer: 29681-45-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR