2-Methyl-3-nitroanisol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR25731-1G
OR25731-5G
OR25731-25G
APOSOR25731-1GEA
218
DKK
APOSOR25731-1G
APOSOR25731-5G
APOSOR25731-25G
2-Methyl-3-nitroanisol 98%
2-Methyl-3-nitroanisol
Formel:
C₈H₉NO₃ Kogepunkt: 94…96 °C/0,5mmHg Smeltepunkt: 54…56 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007159 CAS-nummer: 4837-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...