(±)-Diethyl acetylsuccinate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR25059-100G
OR25059-25G
OR25059-5G
APOSOR25059-100GEA
1520
DKK
APOSOR25059-100G
APOSOR25059-25G
APOSOR25059-5G
(±)-Diethyl acetylsuccinate
(±)-Diethyl acetylsuccinate
Formel:
C₁₀H₁₆O₅ Kogepunkt: 180…183 °C Smeltepunkt: –8 °C Massefylde: 1,08 g/cm³ (25 °C) Flammepunkt: 112 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009157 CAS-nummer: 1115-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...