4-(tert-Butyl)benzenesulphonamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR2445-100G OR2445-25G OR2445-5G
APOSOR2445-100GEA 535 DKK
APOSOR2445-100G APOSOR2445-25G APOSOR2445-5G
4-(tert-Butyl)benzenesulphonamide
4-(tert-Butyl)benzenesulphonamide
Formel: C₁₀H₁₅NO₂S
Smeltepunkt: 138…142 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00068599
CAS-nummer: 6292-59-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR