(Methylsulfonyl)benzen

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR2412-10G OR2412-25G
APOSOR2412-10GEA 164 DKK
APOSOR2412-10G APOSOR2412-25G
(Methylsulfonyl)benzen
(Methylsulfonyl)benzen
Formel: C₇H₈O₂S
Smeltepunkt: 85…87 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00014741
CAS-nummer: 3112-85-4
EINECS: 221-476-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR