2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 2-Hydrazinyl-4,5-dihydro-1H-imidazole hydrobromide, 2-Hydrazino-2-imidazoline Hydrobromide, 2-Hydrazino-4,5-dihydroimidazole Hydrobromide
OR24108-1G OR24108-50MG OR24108-250MG OR24108-500MG
APOSOR24108-1GEA 556 DKK
APOSOR24108-1G APOSOR24108-50MG APOSOR24108-250MG APOSOR24108-500MG
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
Formel: C₃H₉BrN₄
Smeltepunkt: 181…187 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012694
CAS-nummer: 55959-84-7
EINECS: 259-921-3