5-(4-Methylphenyl)cyclohexane-1,3-dione

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-(p-Tolyl)cyclohexane-1,3-dione

OR23987-1G OR23987-250MG OR23987-2.5G OR23987-5G
APOSOR23987-1GEA 2750 DKK
APOSOR23987-1G APOSOR23987-250MG APOSOR23987-2.5G APOSOR23987-5G
5-(4-Methylphenyl)cyclohexane-1,3-dione
5-(4-Methylphenyl)cyclohexane-1,3-dione
Formel: C₁₃H₁₄O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00111717
CAS-nummer: 61888-37-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR