4,4'-Dichlorbenzhydrol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR2394-100G OR2394-25G
APOSOR2394-100GEA 1560 DKK
APOSOR2394-100G APOSOR2394-25G
4,4'-Dichlorbenzhydrol
4,4'-Dichlorbenzhydrol
Formel: C₁₃H₁₀Cl₂O
Smeltepunkt: 91…93 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000629
CAS-nummer: 90-97-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR