Pentamethylbenzensuphonylchlorid, Technical Grade

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Pentamethylbenzene-1-sulphonyl chloride, 2,3,4,5,6-Pentamethylbenzene-1-sulphonyl chloride, Pentamethylbenzenesulphonyl chloride

OR23675-100G OR23675-25G OR23675-5G
APOSOR23675-100GEA 5560 DKK
APOSOR23675-100G APOSOR23675-25G APOSOR23675-5G
Pentamethylbenzensuphonylchlorid, Technical Grade
Pentamethylbenzensuphonylchlorid
Formel: C₁₁H₁₅ClO₂S
Smeltepunkt: 81…85 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00014722
CAS-nummer: 52499-94-2
UN nummer: 3261
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR