Cyclohexylbenzen
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Phenylcyclohexane
OR2340-100G
OR2340-25G
APOSOR2340-100GEA
186
DKK
APOSOR2340-100G
APOSOR2340-25G
Cyclohexylbenzen
Cyclohexylbenzen
Formel:
C₁₂H₁₆ Molekylvægt: 160.26 g/mol Kogepunkt: 239…240 °C Smeltepunkt: 44381 °C Massefylde: 0.95 g/cm³ (25 °C) Flammepunkt: 81 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001451 CAS-nummer: 827-52-1 EINECS: 212-572-0 UN nummer: 3082 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...