1-(3-Bromphenyl)pyrrol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR23352-1G
OR23352-250MG
APOSOR23352-1GEA
430
DKK
APOSOR23352-1G
APOSOR23352-250MG
1-(3-Bromphenyl)pyrrol
1-(3-Bromophenyl)pyrrole
Formel:
C₁₀H₈BrN Smeltepunkt: 64…65 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02665243 CAS-nummer: 107302-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...