5-Phenylisoxazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR23194-1G
OR23194-500MG
OR23194-5G
APOSOR23194-1GEA
382
DKK
APOSOR23194-1G
APOSOR23194-500MG
APOSOR23194-5G
5-Phenylisoxazole
5-Phenylisoxazole
Formel:
C₉H₇NO Smeltepunkt: 23…25 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00090038 CAS-nummer: 1006-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...