2,1,3-Benzothiadiazole-5-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR23124-1G OR23124-5G
APOSOR23124-1GEA 785 DKK
APOSOR23124-1G APOSOR23124-5G
2,1,3-Benzothiadiazole-5-carboxylic acid
2,1,3-Benzothiadiazole-5-carboxylic acid
Formel: C₇H₄N₂O₂S
Smeltepunkt: 230…232 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01647555
CAS-nummer: 16405-98-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR