Benzothiazole-5-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR23085-1G
OR23085-250MG
APOSOR23085-1GEA
644
DKK
APOSOR23085-1G
APOSOR23085-250MG
Benzothiazole-5-carboxylic acid
Benzothiazole-5-carboxylic acid
Formel:
C₈H₅NO₂S Molekylvægt: 179.2 g/mol Smeltepunkt: 258…261 °C (dec.) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06796380 CAS-nummer: 68867-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...