1-Chlorophthalazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR2278-1G OR2278-500MG OR2278-5G
APOSOR2278-1GEA 1860 DKK
APOSOR2278-1G APOSOR2278-500MG APOSOR2278-5G
1-Chlorophthalazine
1-Chlorophthalazine
Formel: C₈H₅ClN₂
Smeltepunkt: 109…112 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00024141
CAS-nummer: 5784-45-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR