Dimethyl aminomalonate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR22760-50MG OR22760-250MG
APOSOR22760-50MGEA 710 DKK
APOSOR22760-50MG APOSOR22760-250MG
Dimethyl aminomalonate
Dimethyl aminomalonate
Formel: C₅H₉NO₄
Molekylvægt: 147.13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00829425
CAS-nummer: 53704-09-9
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR