2-(5-Chloro-2-nitrophenyl)acetonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR22114-1G OR22114-250MG
APOSOR22114-1GEA 2310 DKK
APOSOR22114-1G APOSOR22114-250MG
2-(5-Chloro-2-nitrophenyl)acetonitrile
2-(5-Chloro-2-nitrophenyl)acetonitrile
Formel: C₈H₅ClN₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00114693
CAS-nummer: 72301-65-6
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR