2-Acetyl-1H-pyrrol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR21519-5G
OR21519-100G
APOSOR21519-5GEA
132
DKK
APOSOR21519-5G
APOSOR21519-100G
2-Acetyl-1H-pyrrol 98%
2-Acetyl-1H-pyrrol
Formel:
C₆H₇NO Kogepunkt: 220 °C Smeltepunkt: 88…93 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005220 CAS-nummer: 1072-83-9 EINECS: 214-016-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...