4-Azaxanthone 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR21226-1G OR21226-5G
APOSOR21226-1GEA 0 DKK
APOSOR21226-1G APOSOR21226-5G
4-Azaxanthone 97%
4-Azaxanthone
Formel: C₁₂H₇NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052253
CAS-nummer: 6537-46-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR