3-(2,4-Dinitrophenoxy)benzaldehyde
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR200042-1G
OR200042-100MG
OR200042-250MG
OR200042-500MG
OR200042-2.5G
OR200042-5G
APOSOR200042-1GEA
2650
DKK
APOSOR200042-1G
APOSOR200042-100MG
APOSOR200042-250MG
APOSOR200042-500MG
APOSOR200042-2.5G
APOSOR200042-5G
3-(2,4-Dinitrophenoxy)benzaldehyde
3-(2,4-Dinitrophenoxy)benzaldehyde
Formel:
C₁₃H₈N₂O₆ Smeltepunkt: 123…124 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00413206 CAS-nummer: 2363-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...