(R)-(-)-β-Aminophenethylalkohol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

OR19518-25G OR19518-5G OR19518-100G
APOSOR19518-25GEA 371 DKK
APOSOR19518-25G APOSOR19518-5G APOSOR19518-100G
(R)-(-)-β-Aminophenethylalkohol 97%
(R)-(-)-β-Aminophenethylalkohol
Formel: C₈H₁₁NO
Molekylvægt: 137.18 g/mol
Smeltepunkt: 75…78 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008062
CAS-nummer: 56613-80-0
EINECS: 260-287-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR