1-Methyl-2-indoleboronic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(1-Methyl-1H-indol-2-yl)boronic acid
OR1923-1G
OR1923-250MG
APOSOR1923-1GEA
1080
DKK
APOSOR1923-1G
APOSOR1923-250MG
1-Methyl-2-indoleboronic acid 95%
1-Methyl-2-indoleboronic acid
Formel:
C₉H₁₀BNO₂ Smeltepunkt: 108…112 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD01114668 CAS-nummer: 191162-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...