3-Quinolincarbonitril

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Cyanoquinoline

OR18815-1G OR18815-5G
APOSOR18815-1GEA 251 DKK
APOSOR18815-1G APOSOR18815-5G
3-Quinolincarbonitril
3-Quinolincarbonitril
Formel: C₁₀H₆N₂
Molekylvægt: 154.17
Smeltepunkt: 108…110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006766
CAS-nummer: 34846-64-5
EINECS: 252-248-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR