4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Chloro-7-azaindole-5-carboxylic acid

OR18604-1G OR18604-250MG
APOSOR18604-1GEA 1140 DKK
APOSOR18604-1G APOSOR18604-250MG
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 95%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid
Formel: C₈H₅ClN₂O₂
Molekylvægt: 196.5900 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10574983
CAS-nummer: 920966-03-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR