(Bromomethyl)cyclopentane 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR18584-1G OR18584-5G OR18584-25G
APOSOR18584-1GEA 164 DKK
APOSOR18584-1G APOSOR18584-5G APOSOR18584-25G
(Bromomethyl)cyclopentane 97%
Formel: C₆H₁₁Br
Kogepunkt: 44 °C
Massefylde: 1,271 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09263483
CAS-nummer: 3814-30-0
UN nummer: 1993
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR