2-Iodanilin 98+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR1857-100G
OR1857-25G
APOSOR1857-100GEA
622
DKK
APOSOR1857-100G
APOSOR1857-25G
2-Iodanilin 98+%
2-Iodanilin
Formel:
C₆H₆IN Smeltepunkt: 53…58 °C Flammepunkt: 113 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00007680 CAS-nummer: 615-43-0 EINECS: 210-426-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...