Iminostilbene 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5H-Dibenzo[b,f]azepine
OR18383-100G
OR18383-25G
APOSOR18383-100GEA
600
DKK
APOSOR18383-100G
APOSOR18383-25G
Iminostilbene 97%
Iminostilbene
Formel:
C₁₄H₁₁N Smeltepunkt: 196…199 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005071 CAS-nummer: 256-96-2 EINECS: 205-970-0 UN nummer: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...