2-Chlorbenzimidazol 96%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Chloro-1H-benzo[d]imidazole, 2-Chloro-1H-benzimidazole
OR18361-100G
OR18361-25G
OR18361-5G
APOSOR18361-100GEA
513
DKK
APOSOR18361-100G
APOSOR18361-25G
APOSOR18361-5G
2-Chlorbenzimidazol 96%
2-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ Smeltepunkt: 180…185 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00051944 CAS-nummer: 4857-06-1 EINECS: 225-453-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...