Bis(4-Chlorphenyl)sulfon 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)

OR18307-100G OR18307-500G
APOSOR18307-100GEA 545 DKK
APOSOR18307-100G APOSOR18307-500G
Bis(4-Chlorphenyl)sulfon 99%
Bis(4-Chlorphenyl)sulfon
Formel: C₁₂H₈Cl₂O₂S
Kogepunkt: 249…251 °C/10mmHg
Smeltepunkt: 143…150 °C
Flammepunkt: 233 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000619
CAS-nummer: 80-07-9
EINECS: 201-247-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR