6-Bromoimidazo[1,2-a]pyridin-8-amine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR17942-1G
OR17942-250MG
APOSOR17942-1GEA
183
DKK
APOSOR17942-1G
APOSOR17942-250MG
6-Bromoimidazo[1,2-a]pyridin-8-amine
6-Bromoimidazo[1,2-a]pyridin-8-amine
Formel:
C₇H₆BrN₃ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD13250043 CAS-nummer: 676371-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...