4-Bromo-N,N-diphenylaniline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Bromophenyl)diphenylamine, 4-Bromotriphenylamine

OR17665-100G OR17665-25G
APOSOR17665-100GEA 872 DKK
APOSOR17665-100G APOSOR17665-25G
4-Bromo-N,N-diphenylaniline 98%
4-Bromo-N,N-diphenylaniline
Formel: C₁₈H₁₄BrN
Molekylvægt: 324.21 g/mol
Smeltepunkt: 108…112 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01851266
CAS-nummer: 36809-26-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR