Cyclohexylboronic acid pinacol ester 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR17650-100G OR17650-25G OR17650-5G
APOSOR17650-100GEA 2010 DKK
APOSOR17650-100G APOSOR17650-25G APOSOR17650-5G
Cyclohexylboronic acid pinacol ester 95%
Cyclohexylboronic acid pinacol ester
Formel: C₁₂H₂₃BO₂
Massefylde: 0,93 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04038749
CAS-nummer: 87100-15-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR