4-Bromoisoquinolin-1(2H)-one
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR17550-1G
OR17550-5G
OR17550-25G
APOSOR17550-1GEA
175
DKK
APOSOR17550-1G
APOSOR17550-5G
APOSOR17550-25G
4-Bromoisoquinolin-1(2H)-one
4-Bromoisoquinolin-1(2H)-one
Formel:
C₉H₆BrNO Molekylvægt: 224.0500 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08437036 CAS-nummer: 3951-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...