Benzyloxyacetic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR17515-100G OR17515-25G
APOSOR17515-100GEA 0 DKK
APOSOR17515-100G APOSOR17515-25G
Benzyloxyacetic acid 97%
Benzyloxyacetic acid
Formel: C₉H₁₀O₃
Kogepunkt: 137…139 °C/0,6mmHg
Massefylde: 1,162 g/cm³ (25 °C)
Flammepunkt: 113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00274211
CAS-nummer: 30379-55-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR