2-Chloroquinazoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR17301-1G OR17301-250MG
APOSOR17301-1GEA 305 DKK
APOSOR17301-1G APOSOR17301-250MG
2-Chloroquinazoline
2-Chloroquinazoline
Formel: C₈H₅ClN₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09959715
CAS-nummer: 6141-13-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR